missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl Methyl Ether, Supelco™
Description
LiChrosolv™ solvents provide high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level. This makes them ideal for reproducible separations.
Specifications
Specifications
| Melting Point | -108.6°C |
| Density | 0.74 g/cm3 at 20°C |
| Quantity | 2.5 L |
| Molecular Formula | (CH3)3COCH3 |
| MDL Number | MFCD00008812 |
| Synonym | tert-Butyl methyl ether, MTBE, MTB, 2-Methoxy-2-methylpropane, Methyl tert-butyl ether, TBME, tert-Butyl methyl ether |
| Boiling Point | 55.3°C/1013 hPa |
| Molecular Weight (g/mol) | 88.15 |
| Grade | Research |
| Chemical Name or Material | Tert-Butyl Methyl Ether |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?