missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl ether, For Spectroscopy Uvasol™, MilliporeSigma™

Product Code. 15864160
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15864160 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15864160 Supplier Supelco Supplier No. 1019841000
 Restricted Item
Estimated Shipment: 17-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For spectroscopy Uvasol™

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -108.6°C
CAS 1634-04-4
Color Colorless
Density 0.74g/cm3 at 20°C
Quantity 1 L
Flash Point −28°C (−18.4°F)
Ignition Point 460°C (860°F)
Odor Charactristic
Packaging Glass Bottle
Physical Form Liquid
Molecular Formula C5H12O
Vapor Pressure 268 hPa (20°C)
UN Number 2398
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
Boiling Point 55.3°C
Solubility Information Soluble in water: 42g/L at 20°C
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
SMILES CC(C)(C)OC
IUPAC Name 2-methoxy-2-methylpropane
Molecular Weight (g/mol) 88.15
PubChem CID 15413
ChEBI CHEBI:27642
Formula Weight 88.15
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.