Learn More
tert-Butyl(dec-9-yn-1-yloxy)dimethylsilane, TRC
Specifications
Specifications
| Chemical Name or Material | tert-Butyl(dec-9-yn-1-yloxy)dimethylsilane |
| CAS | 96045-10-2 |
| Molecular Formula | C16H32OSi |
| Quantity | 1 g |
| Synonym | (9-Decyn-1-yloxy)(1,1-dimethylethyl)dimethylsilane, 1-(tert-Butyldimethylsilyloxy)-9-decyne, 10-[(tert-Butyldimethylsilyl)oxy]-1-decyne, 9-Decyn-1-ol tert-Butyldimethylsilyl Ether |
| InChI Formula | InChI=1S/C16H32OSi/c1-7-8-9-10-11-12-13-14-15-17-18(5,6)16(2,3)4/h1H,8-15H2,2-6H3 |
| SMILES | CC(C)(C)[Si](C)(C)OCCCCCCCCC#C |
| IUPAC Name | tert-butyl-dec-9-ynoxy-dimethylsilane |
| Molecular Weight (g/mol) | 268.51 |
| Formula Weight | 268.2222 |