Learn More
tert-Butyl 5-Hydroxypentanoate, TRC
Specifications
Specifications
| Chemical Name or Material | tert-Butyl 5-Hydroxypentanoate |
| CAS | 91420-49-4 |
| Molecular Formula | C9H18O3 |
| Quantity | 50 mg |
| Synonym | 5-Hydroxypentanoic Acid 1,1-Dimethylethyl Ester |
| InChI Formula | InChI=1S/C9H18O3/c1-9(2,3)12-8(11)6-4-5-7-10/h10H,4-7H2,1-3H3 |
| SMILES | CC(C)(C)OC(=O)CCCCO |
| IUPAC Name | tert-butyl 5-hydroxypentanoate |
| Molecular Weight (g/mol) | 174.24 |
| Formula Weight | 174.1256 |