Learn More
tert-Butanol, ACS Reagent, ≥99.0%, Honeywell Riedel-de Haën™
Product Code. 15695080
Change view
Click to view available options
Quantity:
500 mL
1 L
2 L
20 L
Packaging:
Glass Bottle
Poly Bottle
Stainless Steel Can
Unit Size:
1 L
2 L
20 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15695080 2 L Glass Bottle 2 L
15605090 500 mL Glass Bottle 500mL
15665080 1 L Poly Bottle 1 L
15685080 20 L Stainless Steel Can 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15695080 Supplier Honeywell-Riedel-de Haen Supplier No. 3605382L
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
ACS Reagent,≥99.0%
ACS GENERAL USE SOLVENTS
  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004464
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
Synonym tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
PubChem CID 6386
ChEBI CHEBI:45895
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O
Specifications
Melting Point 23°C to 26°C (literature)
CAS Max % 100.0000%
Density 0.775 g/mL at 25 °C(lit.)
Quantity 2 L
Explosion Limit 0.08
Flash Point 11°C
Ignition Point 896°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.387
Linear Formula (CH3)3COH
Vapor Pressure 44mmHg (26°C), 31mmHg (20°C)
UN Number UN1120
Beilstein 906698
Boiling Point 81°C to 83°C
Formula Weight 74.12
Percent Purity >99%
Grade ACS
Chemical Name or Material tert-Butanol, ACS Reagent, ≥99.0%