Learn More
3M™ Nomad™ Terra Heavy Traffic Entrance Matting 8100
Product Code. 17138710
Click to view available options
Dimensions (L x W):
152 x 91.4 cm
610 x 91.4 cm
910 x 91.4 cm
180 x 122 cm
610 x 122 cm
910 x 122 cm
Color:
Black
Brown
Gray
Green
Red
Silver
Unit Size:
1 roll
Each
Product Code. 17138710
Brand: 3M™ Nomad™ 7100116624
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Entrance coiled vinyl scraper matting designed for heavy traffic applications. The construction allows dirt and moisture to flow through the matting so it is not retracked into the building. Grease and oil resistant.
  • Unbacked scraper matting designed for high traffic applications
  • High quality vinyl offers excellent crush resistance
  • Unbacked, open construction of the mat allows water to drain and soil to pass through, out of sight
  • Easily vacuum out trapped dirt
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W) 152 x 91.4 cm
For Use With (Application) Heavy traffic areas
Model Nomad™ Terra Dirt Control Mats
Thickness (Metric) 17 mm
Color Gray
Length (Metric) 152 cm
Material Elastic Vinyl
Width (Metric) 91.4 cm