Learn More
Terfenadine, Mikromol™
Description
Terfenadine
Specifications
Specifications
| Chemical Name or Material | Terfenadine |
| CAS | 50679-08-8 |
| Analyte or Component Names | Terfenadine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C32 H41 N O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Terfenadine |
| InChI Formula | InChI=1S/C32H41NO2/c1-31(2,3)26-18-16-25(17-19-26)30(34)15-10-22-33-23-20-29(21-24-33)32(35,27-11-6-4-7-12-27)28-13-8-5-9-14-28/h4-9,11-14,16-19,29-30,34-35H,10,15,20-24H2,1-3H3 |
| SMILES | CC(C)(C)c1ccc(cc1)C(O)CCCN2CCC(CC2)C(O)(c3ccccc3)c4ccccc4 |
| IUPAC Name | 1-(4-tert-butylphenyl)-4-[4-[hydroxy(diphenyl)methyl]piperidin-1-yl]butan-1-ol |
| Show More |