Learn More
Terbuthylazine, TRC
Product Code. 17950895
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17950895 1 g 1g
17914355 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17950895 Supplier Toronto Research Chemicals Supplier No. TRCT1105001G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5915-41-3
Molecular Formula C9 H16 Cl N5
Molecular Weight (g/mol) 229.71
Synonym Terbuthylazine, 1,3,5-Triazine-2,4-diamine, 6-chloro-N2-(1,1-dimethylethyl)-N4-ethyl- (ACI), 1,3,5-Triazine-2,4-diamine, 6-chloro-N-(1,1-dimethylethyl)-N'-ethyl- (9CI), 6-Chloro-N2-(1,1-dimethylethyl)-N4-ethyl-1,3,5-triazine-2,4-diamine (ACI), s-Triazine, 2-(tert-butylamino)-4-chloro-6-(ethylamino)- (7CI, 8CI), 2-Chloro-4-(ethylamino)-6-(tert-butylamino)-s-triazine, 2-Chloro-4-ethylamino-6-tert-butylamino-1,3,5-triazine, 2-Chloro-4-tert-butylamino-6-ethylamino-s-triazine, 2-N-tert-Butyl-6-chloro-4-N-ethyl-1,3,5-triazine-2,4-diamine, 2-tert-Butylamino-4-chloro-6-ethylamino-s-triazine, 4-Ethylamino-6-tert-butylamino-2-chloro-S-triazine, 6-Chloro-2-tert-butylamino-4-(ethylamino)-[1,3,5]-triazine, ChlorCaragard, Click, Click 500SC, G 13529, Gardeprim A 1862, Gardoprim, GS 13529, N-tert-Butyl-6-chloro-N'-ethyl-1,3,5-triazine-2,4-diamine, Primatol M, Terbutazine, Terbutylazine, Terbutylethylazine, tert-Butylazine, Tyllanex
IUPAC Name 2-N-tert-butyl-6-chloro-4-N-ethyl-1,3,5-triazine-2,4-diamine
SMILES CCNc1nc(Cl)nc(NC(C)(C)C)n1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C9H16ClN5/c1-5-11-7-12-6(10)13-8(14-7)15-9(2,3)4/h5H2,1-4H3,(H2,11,12,13,14,15)
Formula Weight 229.1094
Chemical Name or Material Terbuthylazine