missing translation for 'onlineSavingsMsg'
Learn More

Terbuthylazine, MedChemExpress

Product Code. 16408826
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16408826 100 mg 100mg
16402296 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16408826 Supplier Medchem Express Supplier No. HYB1847
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Terbuthylazine is an inhibitor of acetolactate syntase (ALS), is a selective herbicide.

Terbuthylazine is an inhibitor of acetolactate syntase (ALS), is a selective herbicide.

Specifications

Chemical Name or Material Terbuthylazine
CAS 5915-41-3
Color White
Purity Grade Notes Research
Molecular Formula C9H16ClN5
Quantity 100 mg
Solubility Information DMSO : 23 mg/mL (100.13 mM; Need ultrasonic and warming)
SMILES CCNC1=NC(Cl)=NC(NC(C)(C)C)=N1
Molecular Weight (g/mol) 229.71
Formula Weight 229.71
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.