Learn More
Terazosin (piperazine D8), TRC
Specifications
Specifications
| Chemical Name or Material | Terazosin (piperazine D8) |
| CAS | 1006718-20-2 |
| Molecular Formula | C19H17D8N5O4 |
| Quantity | 2.5 mg |
| Synonym | 1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-[(tetrahydro-2-furanyl)carbonxyl]piperazine-d8 |
| InChI Formula | InChI=1S/C19H25N5O4/c1-26-15-10-12-13(11-16(15)27-2)21-19(22-17(12)20)24-7-5-23(6-8-24)18(25)14-4-3-9-28-14/h10-11,14H,3-9H2,1-2H3,(H2,20,21,22)/i5D2,6D2,7D2,8D2 |
| SMILES | NC1=NC(N2C([2H])([2H])C([2H])([2H])N(C(C3CCCO3)=O)C([2H])([2H])C2([2H])[2H])=NC4=CC(OC)=C(OC)C=C41 |
| Molecular Weight (g/mol) | 395.48 |
| Formula Weight | 182.079 |