missing translation for 'onlineSavingsMsg'
Learn More

Tenofovir Alafenamide Fumarate, TRC

Product Code. 30136331
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136331 500 mg 500mg
30133410 25 mg 25mg
30012130 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136331 Supplier Toronto Research Chemicals Supplier No. TRCT779380500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 379270-38-9
Molecular Formula C21 H29 N6 O5 P . C4 H4 O4
Molecular Weight (g/mol) 592.5381
Synonym N-[(S)-[[(1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phenoxyphosphinyl]-L-alanine 1-Methylethyl Ester (2E)-2-butenedioate (1:1), GS 7340 Fumarate
IUPAC Name (E)-but-2-enedioic acid;propan-2-yl (2S)-2-[[[(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethyl-phenoxyphosphoryl]amino]propanoate
SMILES CC(C)OC(=O)[C@H](C)N[P@@](=O)(CO[C@H](C)Cn1cnc2c(N)ncnc12)Oc3ccccc3.OC(=O)\C=C\C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C21H29N6O5P.C4H4O4/c1-14(2)31-21(28)16(4)26-33(29,32-17-8-6-5-7-9-17)13-30-15(3)10-27-12-25-18-19(22)23-11-24-20(18)27;5-3(6)1-2-4(7)8/h5-9,11-12,14-16H,10,13H2,1-4H3,(H,26,29)(H2,22,23,24);1-2H,(H,5,6)(H,7,8)/b;2-1+/t15-,16+,33-;/m1./s1
Formula Weight 592.2047
Percent Purity >95%
Chemical Name or Material Tenofovir Alafenamide Fumarate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.