Learn More
Tenofovir Alafenamide Fumarate, TRC
Product Code. 30133410
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133410 25 mg 25mg
30136331 500 mg 500mg
30012130 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133410 Supplier Toronto Research Chemicals Supplier No. TRCT77938025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 379270-38-9
Molecular Formula C21 H29 N6 O5 P . C4 H4 O4
Molecular Weight (g/mol) 592.5381
Synonym N-[(S)-[[(1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phenoxyphosphinyl]-L-alanine 1-Methylethyl Ester (2E)-2-butenedioate (1:1), GS 7340 Fumarate
IUPAC Name (E)-but-2-enedioic acid;propan-2-yl (2S)-2-[[[(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethyl-phenoxyphosphoryl]amino]propanoate
SMILES CC(C)OC(=O)[C@H](C)N[P@@](=O)(CO[C@H](C)Cn1cnc2c(N)ncnc12)Oc3ccccc3.OC(=O)\C=C\C(=O)O
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C21H29N6O5P.C4H4O4/c1-14(2)31-21(28)16(4)26-33(29,32-17-8-6-5-7-9-17)13-30-15(3)10-27-12-25-18-19(22)23-11-24-20(18)27;5-3(6)1-2-4(7)8/h5-9,11-12,14-16H,10,13H2,1-4H3,(H,26,29)(H2,22,23,24);1-2H,(H,5,6)(H,7,8)/b;2-1+/t15-,16+,33-;/m1./s1
Formula Weight 592.2047
Percent Purity >95%
Chemical Name or Material Tenofovir Alafenamide Fumarate
Grade HPLC