Learn More
Tenofovir alafenamide fumarate, MedChemExpress
Product Code. 16496676
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496676 100 mg 100mg
16496378 10 mg 10mg
16488967 5 mg 5mg
16402497 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16496676 Supplier Medchem Express Supplier No. HY15232A
MedChemExpress Tenofovir alafenamide fumarate (GS-7340 fumarate) is an investigational oral prodrug of Tenofovir. Tenofovir is a HIV-1 nucleotide reverse transcriptase inhibitor.
Tenofovir alafenamide fumarate (GS-7340 fumarate) is an investigational oral prodrug of Tenofovir. Tenofovir is a HIV-1 nucleotide reverse transcriptase inhibitor.
Specifications
Chemical Name or Material Tenofovir alafenamide fumarate
CAS 379270-38-9
Color White
Purity Grade Notes Research
Molecular Formula C25H33N6O9P
Quantity 100 mg
Synonym GS-7340 (fumarate)
Solubility Information DMSO : ≥ 36 mg/mL (60.76 mM) ∣H2O : ≥ 25 mg/mL (42.19 mM)
SMILES NC1=NC=NC2=C1N=CN2C[C@@H](C)OC[P@](OC3=CC=CC=C3)(N[C@@H](C)C(OC(C)C)=O)=O.O=C(O)/C=C/C(O)=O
Molecular Weight (g/mol) 592.54
Formula Weight 592.54
Percent Purity 99.14%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus