Learn More
Teniposide, MedChemExpress
Product Code. 16446316
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16446316 100 mg 100mg
16443386 25 mg 25mg
16421997 50 mg 50mg
16421106 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16446316 Supplier Medchem Express Supplier No. HY13761
 Restricted Item
Estimated Shipment: 30-11-2026
MedChemExpress Teniposide is a podophyllotoxin derivative, acts as a topoisomerase II inhibitor, and used as a chemotherapeutic agent.
Teniposide is a podophyllotoxin derivative, acts as a topoisomerase II inhibitor, and used as a chemotherapeutic agent.
Specifications
Chemical Name or Material Teniposide
CAS 29767-20-2
Color White
Purity Grade Notes Research
Molecular Formula C32H32O13S
Quantity 100 mg
Synonym VM26
Solubility Information DMSO : ≥ 30 mg/mL (45.69 mM)
SMILES O=C1OC[C@]2([H])[C@H](O[C@H]3[C@@H]([C@H]([C@@H]([C@@H](CO4)O3)O[C@@H]4C5=CC=CS5)O)O)C6=C(C=C7OCOC7=C6)[C@@H](C8=CC(OC)=C(O)C(OC)=C8)[C@]21[H]
Molecular Weight (g/mol) 656.65
Formula Weight 656.65
Percent Purity 97.46%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death