missing translation for 'onlineSavingsMsg'
Learn More

Teneligliptin hydrobromide, MedChemExpress

Product Code. 16496178
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496178 10 mg 10mg
16462317 50 mg 50mg
16446556 100 mg 100mg
16419865 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496178 Supplier Medchem Express Supplier No. HY14806A
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Teneligliptin (MP-513) hydrobromide is a potent chemotype prolylthiazolidine-based DPP-4 inhibitor, which competitively inhibits human plasma, rat plasma, and human recombinant DPP-4 in vitro, with IC50s of approximately 1 nM.

Teneligliptin (MP-513) hydrobromide is a potent chemotype prolylthiazolidine-based DPP-4 inhibitor, which competitively inhibits human plasma, rat plasma, and human recombinant DPP-4 in vitro, with IC50s of approximately 1 nM.

Specifications

Chemical Name or Material Teneligliptin hydrobromide
CAS 906093-29-6
Color Colorless
Purity Grade Notes Research
Molecular Formula C22H30N6OS.5/2HBr
Quantity 10 mg
Synonym MP-513 hydrobromide
Solubility Information H2O : ≥ 200 mg/mL (318.04 mM) ∣DMSO : ≥ 100 mg/mL (159.02 mM)
SMILES O=C(N1CSCC1)[C@H]2NC[C@@H](N3CCN(C4=CC(C)=NN4C5=CC=CC=C5)CC3)C2.[2.5 HBr]
Molecular Weight (g/mol) 628.86
Formula Weight 628.86
Percent Purity 98.71%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.