missing translation for 'onlineSavingsMsg'
Learn More

Teneligliptin Hydrobromide (2:5), TRC

Product Code. 30417040
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417040 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417040 Supplier Toronto Research Chemicals Supplier No. TRCT018300100MG
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Teneligliptin Hydrobromide (2:5)
CAS 906093-29-6
Molecular Formula C22H32.5Br2.5N6OS
Quantity 100 mg
Synonym [(2S,4S)-4-[4-(3-Methyl-1-phenyl-1H-pyrazol-5-yl)-1-piperazinyl]-2-pyrrolidinyl]-3-thiazolidinylmethanone Hydrobromide (2:5), 3-[[(2S,4S)-4-[4-(3-Methyl-1-phenyl-1H-pyrazol-5-yl)-1-piperazinyl]-2-pyrrolidinyl]carbonyl]thiazolidine Hydrobromide (2:5), Tenelia Hydrobromide (2:5)
InChI Formula InChI=1S/2C22H30N6OS.5BrH/c2*1-17-13-21(28(24-17)18-5-3-2-4-6-18)26-9-7-25(8-10-26)19-14-20(23-15-19)22(29)27-11-12-30-16-27;;;;;/h2*2-6,13,19-20,23H,7-12,14-16H2,1H3;5*1H/t2*19-,20-;;;;;/m00...../s1
SMILES O=C(N1CSCC1)[C@H]2NC[C@@H](N(CC3)CCN3C4=CC(C)=NN4C5=CC=CC=C5)C2
IUPAC Name [(2S,4S)-4-[4-(5-methyl-2-phenylpyrazol-3-yl)piperazin-1-yl]pyrrolidin-2-yl]-(1,3-thiazolidin-3-yl)methanone;pentahydrobromide
Molecular Weight (g/mol) 628.86
Formula Weight 1252.0712
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.