missing translation for 'onlineSavingsMsg'
Learn More

Temsavir, MedChemExpress

Product Code. 16486518
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486518 10 mg 10mg
16476766 100 mg 100mg
16442617 50 mg 50mg
16429107 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16486518 Supplier Medchem Express Supplier No. HY15440

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Temsavir (BMS-626529) is a novel attachment inhibitor that targets HIV-1 gp120 and prevents its binding to CD4+ T cells.

Temsavir (BMS-626529) is a novel attachment inhibitor that targets HIV-1 gp120 and prevents its binding to CD4+ T cells.

Specifications

Chemical Name or Material Temsavir
CAS 701213-36-7
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C24H23N7O4
Quantity 10 mg
Synonym BMS-626529
Solubility Information DMSO : ≥ 16.67 mg/mL (35.21 mM)
SMILES O=C(N1CCN(C(C2=CC=CC=C2)=O)CC1)C(C3=CNC4=C3C(OC)=CN=C4N5C=NC(C)=N5)=O
Molecular Weight (g/mol) 473.48
Formula Weight 473.48
Percent Purity 98.11%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.