missing translation for 'onlineSavingsMsg'
Learn More

Tempo, MP Biomedicals

Product Code. 11498012 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498012 1 g 1g
11408022 5 g 5g
11418022 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498012 Supplier MP Biomedicals Supplier No. 0215268101
  • ONLINE EXCLUSIVE PRICE
    57.35 EUR
    54.60 EUR / 1g
    Save 2.75 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2564-83-2 | Formula: C9H18NO | Weight: 156.25 g/mol

Chemical Identifiers

CAS 2564-83-2
Molecular Formula C9H18NO
Molecular Weight (g/mol) 156.25
MDL Number MFCD00009599
InChI Key QYTDEUPAUMOIOP-UHFFFAOYSA-N
IUPAC Name (2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
SMILES CC1(C)CCCC(C)(C)N1[O]

Specifications

Quantity 1 g
Formula Weight 156.2
Physical Form Solid
Chemical Name or Material Tempo
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.