Description
- Use with a temperature range 0 to 180°C
- Includes support plate and quick release closure NW 60
Specifications
Specifications
| Product Type | Temperature Stabilization Jacket |
| For Use With (Equipment) | Viscometer |
| For Use With (Application) | Viscometer |
| Temperature Range (Metric) | 0°C to 180°C |