Learn More
Thermo Scientific™ Temperature Chart Recorder Paper
Product Code. 10354557 Shop All Thermo Scientific Products
Change view
Click to view available options
Temperature Range:
-200°C to 0°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range Unit Size
10354557 -200°C to 0°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10354557 Supplier Thermo Scientific™ Supplier No. 7289
  • ONLINE EXCLUSIVE PRICE
    112.00 EUR
    109.00 EUR / Pack of 1
    Save 3.00 EUR
    2% Off
 Restricted Item
In Stock
Use Thermo Scientific™ Temperature Chart Recorder Paper in seven-day circular temperature chart recorders.
Use Thermo Scientific™ Temperature Chart Recorder Paper in seven-day circular temperature chart recorders.
  • 6' (15.2cm) diameter
  • For use in freestanding and panel-mounted chart recorders
TRUSTED_SUSTAINABILITY
Specifications
Type Chart Recorder Paper
Temperature Range -200°C to 0°C
Style Freestanding
For Use With (Equipment) Free-Standing and Panel-Mounted Chart Recorders
Monitoring Options Recorder
Static Holding Time 7 Days
Diameter (English) 6 in.
Diameter (Metric) 15.2 cm