Learn More
Technical Buffer for pH DKD Certified pH 7.00, METTLER TOLEDO™
Product Code. 10709883
Change view
Click to view available options
Quantity:
250 mL
Packaging:
1Each
6x 250 mL
Unit Size:
1500mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10709883 250 mL 1Each 250mL
10711955 250 mL 6x 250 mL 1500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10709883 Supplier Mettler Toledo Online Supplier No. 51350034
  • ONLINE EXCLUSIVE PRICE
    30.00 EUR
    22.93 EUR / 250mL
    Save 7.07 EUR
    23% Off
 Restricted Item
In Stock
The DKD (Deutscher Kalibrierdienst, German Calibration Service) certified buffers are the perfect product for regulated industries.
  • pH 7.00
TRUSTED_SUSTAINABILITY
Specifications
pH 7.00
Quantity 250 mL
Packaging 1Each