Learn More
Tebufenozide-d9, TRC
Specifications
Specifications
| Chemical Name or Material | Tebufenozide-d9 |
| CAS | 2469006-89-9 |
| Molecular Formula | C22 D9 H19 N2 O2 |
| Quantity | 5 mg |
| Synonym | N'-(4-Ethylbenzoyl)-N-(2-methylpropan-2-yl)-3,5-bis(trideuteriomethyl)-2,4,6-trideuteriobenzohydrazide, 3,5-Di(methyl-d3)benzoic-2,4,6-d3 acid 1-(1,1-dimethylethyl)-2-(4-ethylbenzoyl)hydrazide, 1-tert-Butyl-1-(3,5-dimethylbenzoyl-d9)-2-(4-ethylbenzoyl)hydrazine, 3,5-Dimethylbenzoic-d9 acid N-tert-butyl-N-(4-ethylbenzoyl)hydrazide, Tebufenozide-d9, Tebufenozide D9 (3,5-Dimethylbenzoyl-D9) |
| InChI Formula | InChI=1S/C22H28N2O2/c1-7-17-8-10-18(11-9-17)20(25)23-24(22(4,5)6)21(26)19-13-15(2)12-16(3)14-19/h8-14H,7H2,1-6H3,(H,23,25)/i2D3,3D3,12D,13D,14D |
| SMILES | [2H]c1c(C(=O)N(NC(=O)c2ccc(CC)cc2)C(C)(C)C)c([2H])c(c([2H])c1C([2H])([2H])[2H])C([2H])([2H])[2H] |
| IUPAC Name | N-tert-butyl-2,4,6-trideuterio-N'-(4-ethylbenzoyl)-3,5-bis(trideuteriomethyl)benzohydrazide |
| Molecular Weight (g/mol) | 361.525 |
| Formula Weight | 361.272 |