missing translation for 'onlineSavingsMsg'
Learn More

TCF-3/E2A Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18438383 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18438383 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18438383 Supplier Novus Biologicals Supplier No. NBP177119AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TCF-3/E2A Polyclonal antibody specifically detects TCF-3/E2A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCF-3/E2A
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias BHLHB21, bHLHb21TCF-3, Class B basic helix-loop-helix protein 21, E2A immunoglobulin enhancer-binding factor E12/E47, E2AKappa-E2-binding factor, Immunoglobulin enhancer-binding factor E12/E47, Immunoglobulin transcription factor 1, ITF1Transcription factor ITF-1, MGC129647, MGC129648, Transcription factor 3, transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47), transcription factor E2-alpha, VDIR, VDR interacting repressor
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human TCF3. The immunogen is located within amino acids 150 - 200 of TCF3. Amino Acid Squence: LDTQPKKVRKVPPGLPS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6929
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.