Learn More
Tocris Bioscience™ TC SL C5
GREENER_CHOICE
Product Code. 18759306
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18759306
Brand: Tocris Bioscience™ 6884/10
Metastasis-associated lung adenocarcinoma transcript 1 (Malat1) RNA inhibitor
TC SL C5 is a metastasis-associated lung adenocarcinoma transcript 1 (Malat1) RNA inhibitor (KD = 2.9 μM). Binds Malat1 3' terminal stability element for nuclear expression (ENE) triplex. Reduces Malat1 RNA levels by 54% and branching morphogenesis by 38% in mammary tumor organoids. Modulates Malat1 downstream genes without affecting Neat1.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 mg
Molecular Formula C19H21N3O2
Formula Weight 323.4
Product Type Inhibitor
Purity 98%