missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TC JL 37

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
TC JL 37 is a potent inhibitor of tyrosine kinase 2 (TYK2, Ki = 1.6 nM). Shows some selectivity for TYK2 over JAK1/2/3. Inhibits IFNγ production in vivo in mice. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-4-cyano-6-fluoro-N-[2-[[[(1R,2R)-2-fluorocyclopropyl]carbonyl]amino]-4-pyridinyl]benzamide |
| CAS | 1258294-34-6 |
| Assay Percent Range | >98% |
| Quantity | 50 mg |
| Molecular Formula | C17H11ClF2N4O2 |
| Formula Weight | Observed MW: 376.74 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction