missing translation for 'onlineSavingsMsg'
Learn More

Tavaborole, TRC

Product Code. 30423520
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423520 2.5 g 2.5g
30422576 500 mg -
30417658 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423520 Supplier Toronto Research Chemicals Supplier No. TRCT0097652.5G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174671-46-6
Molecular Formula C7 H6 B F O2
Molecular Weight (g/mol) 151.93
Synonym 2,1-Benzoxaborole, 5-fluoro-1,3-dihydro-1-hydroxy-, 5-Fluoro-1,3-dihydro-1-hydroxy-2,1-benzoxaborole, 5-Fluoro-1,3-dihydro-2,1-benzoxaborol-1-ol, AN 2690, AN 2729, Kerydin, Tavaborole
IUPAC Name 5-fluoro-1-hydroxy-3H-2,1-benzoxaborole
SMILES OB1OCc2cc(F)ccc12

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C7H6BFO2/c9-6-1-2-7-5(3-6)4-11-8(7)10/h1-3,10H,4H2
Formula Weight 152.0445
Chemical Name or Material Tavaborole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.