missing translation for 'onlineSavingsMsg'
Learn More

Taurultam, TRC

Product Code. 30412813
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30412813 100 mg
30413051 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30412813 Supplier Toronto Research Chemicals Supplier No. TRCT009600100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38668-01-8
Molecular Formula C3 H8 N2 O2 S
Molecular Weight (g/mol) 136.17
Synonym Tetrahydro-2H-1,2,4-thiadiazine 1,1-Dioxide, ¡1,1-Dioxoperhydro-1,2,4-thiadiazine, Perhydro-1,2,4-thiadiazine 1,1-Dioxide, Tetrahydro-2H-1,2,4-thiadiazine 1,1-Dioxide
IUPAC Name 1,2,4-thiadiazinane 1,1-dioxide
SMILES O=S1(=O)CCNCN1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C3H8N2O2S/c6-8(7)2-1-4-3-5-8/h4-5H,1-3H2
Formula Weight 136.0306
Percent Purity >95%
Chemical Name or Material Taurultam
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.