missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Taurolithocholic Acid-d5 Sodium Salt (Major), TRC
Specifications
Specifications
| Chemical Name or Material | Taurolithocholic Acid-d5 Sodium Salt |
| CAS | 1265476-97-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C26H39D5NNaO5S |
| Quantity | 1 mg |
| Synonym | 2-[[(3α,5β)-3-Hydroxy-24-oxocholan-24-yl-2,2,3,4,4-d5]amino]ethanesulfonic acid sodium salt, Taurolithocholic acid-d5 sodium salt, Taurolithocholic acid D5 sodium salt (3α-hydroxy-5β-cholane-2,2,3,4,4-D5), 2-[[(3α,5β)-3-Hydroxy-24-oxo-2,2,3,4,4-d5-cholan-24-yl]amino]ethanesulfonic acid sodium salt, 3α-Hydroxy-2,2,3,4,4-d5-5β-cholanoyltaurine sodium, 3α-Hydroxy-N-(2-sulfoethyl)-5β-cholan-24-amide-2,2,3,4,4-d5 sodium salt, Lithocholyltaurine-d5 sodium |
| InChI Formula | InChI=1S/C26H45NO5S.Na/c1-17(4-9-24(29)27-14-15-33(30,31)32)21-7-8-22-20-6-5-18-16-19(28)10-12-25(18,2)23(20)11-13-26(21,22)3;/h17-23,28H,4-16H2,1-3H3,(H,27,29)(H,30,31,32);/q;+1/p-1/t17-,18-,19-,20+,21-,22+,23+,25+,26-;/m1./s1/i10D2,16D2,19D; |
| SMILES | [Na+].[2H]C1([2H])C[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC[C@@H]2C([2H])([2H])[C@]1([2H])O)[C@H](C)CCC(=O)NCCS(=O)(=O)[O-] |
| IUPAC Name | sodium;2-[[(4R)-4-[(3R,5R,8R,9S,10S,13R,14S,17R)-2,2,3,4,4-pentadeuterio-3-hydroxy-10,13-dimethyl-1,5,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate |
| Molecular Weight (g/mol) | 510.72 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?