missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Tat-Beclin 1 L11S Peptide - Scrambled Control

Product Code. 18628816 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
1 mg
5 mg
Unit Size:
0.1mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 18628816

Brand: Novus Biologicals™ NBP249887SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Scrambled Control

TRUSTED_SUSTAINABILITY

Specifications

Species Non-species Specific
Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Gene Alias Autophagy Inducing peptide, Tat-Beclin, Tat-Beclin 1, Tat-Beclin 1 peptide, Tat-Beclin peptide, Tat-L11S
Molecular Weight (g/mol) MolecularWeight-theroretical: 3.08 kDa
Quantity 0.1 mg
Research Category Autophagy
Storage Buffer This product is supplied lyophilized. Purity is ≥ to 97% (HPLC)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.