Learn More
Applied Biosystems™ TaqMan™ Array Rat Endogenous Control Panel
Product Code. 11620109 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
2 Cards
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11620109 2 Cards Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11620109 Supplier Applied Biosystems™ Supplier No. 4378704
  • ONLINE EXCLUSIVE PRICE
    1066.00 EUR
    692.90 EUR / Pack of 1
    Save 373.10 EUR
    35% Off
 Restricted Item
Estimated Shipment: 04-12-2026
TaqMan™ Array Rat Endogenous Control Panel
The Applied Biosystems™ TaqMan™ Rat Endogenous Control Array is a 384-well micro flluidic card containing 16 rat TaqMan™ Gene Expression Assays commonly used as endogenous controls arrayed in triplicates. This product is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System. Endogenous Control Panels are sold in packs of 2.
The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
TRUSTED_SUSTAINABILITY
Specifications
Type TaqMan™ Array Rat Endogenous Control Panel
Format 384-well Microfluidic Card
Content And Storage 4°C
For Use With (Application) Pharma and biopharma, real-time PCR (qPCR), real-time PCR-based gene expression profiling, target and lead identification and validation
Includes Control primer-probe set: TaqMan™ rat endogenous control array is sold in a pack of 2 cards and include a CD with assay information and an instrument software input file
Quantity 2 Cards