Learn More
Applied Biosystems™ TaqMan™ Array Human MicroRNA A+B Cards Set v3.0
Shop All Applied Biosystems ProductsDescription
The TaqMan™ Array Human MicroRNA Card Set v3.0 is a two card set containing a total of 384 TaqMan™ MicroRNA Assays per card. The set enables accurate quantitation of 754 human microRNAs in as little as 5 hours. Included on each array is three TaqMan MicroRNA Assay endogenous controls to aid in data normalization and one TaqMan™ MicroRNA Assay not related to human as a negative control. Details on the included assays is available in the Target List file. Purchase of the Megaplex™ RT Primers, Human Pool Set v3.0 is required to run this array set. An additional preamplification step is enabled by Megaplex™ PreAmp Primers, Human Pool Set v3.0 for situations where sensitivity is of the utmost importance and/or sample is limiting.
Specifications
Specifications
| Type | Array Human MicroRNA A+B Cards Set |
| Format | 384-well Microfluidic Card |
| Species | Human |
| Content And Storage | 4°C |
| Form | Dried Down |
| For Use With (Equipment) | AB 7900 HT 384-Well System |
| Label or Dye | FAM |
| Product Line | Megaplex |
| Quantity | 8 pack |
| Reaction Speed | Standard |
| Show More |