Learn More
Applied Biosystems™ TaqMan™ Array Human Angiogenesis Panel
Product Code. 11680109
Change view
Click to view available options
Quantity:
4 Cards
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11680109 4 Cards Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11680109 Supplier Applied Biosystems™ Supplier No. 4378710
  • ONLINE EXCLUSIVE PRICE
    2140.00 EUR
    1391.00 EUR / Pack of 1
    Save 749.00 EUR
    35% Off
 Restricted Item
Estimated Shipment: 20-11-2026
TaqMan™ Array Human Angiogenesis Panel
The Applied Biosystems™ TaqMan™ Human Angiogenesis Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of angiogenesis genes important in drug discovery. This product is designed with 96 TaqMan™ Gene Expression Assays arrayed in 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System and is sold in packs of 4.
This TaqMan™ Gene Signature Array contains 94 assays and two controls to study angiogenesis and lymphangiogenesis in humans. It includes known angiogenesis growth factors like VEGF, and known matrix derived inhibitors such as endostatin. Additionally, the panel contains markers and targets for angiogenesis and lymphangiogenesis. The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
TRUSTED_SUSTAINABILITY
Specifications
Type TaqMan™ Array Human Angiogenesis Panel
Format 384-well Microfluidic Card
Content And Storage 4°C
For Use With (Application) Cell viability, proliferation and function, oncology and genetic disease research, pharma and biopharma, real-time PCR (qPCR), target and lead identification and validation
Includes TaqMan™ array well contains a single TaqMan™ assay for one 2μL reaction
Quantity 4 Cards