Learn More
Gilac™ Tampico Oven Broom
Product Code. 17928461
Change view
Click to view available options
Width (Metric):
38 cm
50 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) unitSize
17928461 38 cm Each
17988631 50 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17928461 Supplier Gilac™ Supplier No. G020020
  • ONLINE EXCLUSIVE PRICE
    65.21 EUR
    47.22 EUR / Pack of 1
    Save 17.99 EUR
    27% Off
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
Tampico Oven Broom is a special brush for bakery and pastry.
  • Special brush for bakery and pastry.
Specifications
Product Type Broom Head
Color Code Orange
Sterility Non-sterile
Autoclavable Not Autoclavable
Width (Metric) 38 cm
Width (English) 15 in.
Color-coded Yes
Material Wood