Learn More
Taltirelin, MedChemExpress
Product Code. 16494407
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494407 50 mg 50mg
16471018 5 mg 5mg
16438548 10 mg 10mg
16438386 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16494407 Supplier Medchem Express Supplier No. HYB0596
 Restricted Item
Estimated Shipment: 02-11-2026
MedChemExpress Taltirelin (TA0910) is a superagonist at thyrotropin-releasing hormone receptor (TRH-R) with an IC50 of 910 nM and EC50 of 36 nM for stimulating an increase in cytosolic Ca2+ concentration (Ca2+ release).
Taltirelin (TA0910) is a superagonist at thyrotropin-releasing hormone receptor (TRH-R) with an IC50 of 910 nM and EC50 of 36 nM for stimulating an increase in cytosolic Ca2+ concentration (Ca2+ release).
Specifications
Chemical Name or Material Taltirelin
CAS 103300-74-9
Color White
Purity Grade Notes Research
Molecular Formula C17H23N7O5
Quantity 50 mg
Synonym TA-0910
Solubility Information DMSO : 100 mg/mL (246.66 mM; Need ultrasonic) ∣H2O : 100 mg/mL (246.66 mM; Need ultrasonic)
SMILES O=C(N(CCC1)[C@@H]1C(N)=O)[C@@H](NC([C@@H](NC2=O)CC(N2C)=O)=O)CC3=CN=CN3
Molecular Weight (g/mol) 405.41
Formula Weight 405.41
Percent Purity 99.67%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation