Learn More
Koettermann™ Tall Storage Cabinets
Product Code. 14855311
Change view
Click to view available options
Depth (Metric):
500 mm
516 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) Unit Size
14855311 516 mm Each
14885291 500 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 14855311 Supplier Koettermann™ Supplier No. 30100084
  • ONLINE EXCLUSIVE PRICE
    1530.00 EUR
    1116.00 EUR / Pack of 1
    Save 414.00 EUR
    27% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-12-2026
Specifications
Product Type Tall Storage Cabinet
Cabinet Type Tall Storage
Depth (Metric) 516 mm
Dimensions (D x W x H) 516 x 900 x 1920 mm
Height (Metric) 192 cm
No. of Doors 2
Width (Metric) 900 mm