Learn More
Koettermann™ Tall Cabinet/Pull-Out Door, 300x1920x516mm
Product Code. 15865115
Change view
Click to view available options
Dimensions (D x W x H):
516 x 300 x 1920 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (D x W x H) Unit Size
15865115 516 x 300 x 1920 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15865115 Supplier Koettermann™ Supplier No. 30100089
  • ONLINE EXCLUSIVE PRICE
    1785.00 EUR
    1549.00 EUR / Pack of 1
    Save 236.00 EUR
    13% Off
Estimated Shipment: 26-11-2026
Koettermann™ Tall Cabinet/Pull-Out Drawer, 300x1920x516mm provides suitable storage to accommodate a variety of laboratory needs.
  • Tall storage cabinet
  • Pull-out drawer
  • Basket set
  • Lockable
Specifications
Dimensions (D x W x H) 516 x 300 x 1920 mm
Includes Pull-out drawer, basket set