missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAFA4/FAM19A4 Mouse anti-Human, Alexa Fluor 750, Clone: 480103, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC5099S-100UG
This item is not returnable.
View return policy
Description
TAFA4/FAM19A4 Monoclonal specifically detects TAFA4/FAM19A4 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| TAFA4/FAM19A4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q96LR4 | |
| FAM19A4 | |
| E. coli-derived recombinant human TAFA4 Ser35-Arg140 Accession # Q96LR4 | |
| RUO | |
| 151647 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 480103 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Chemokine-like protein TAFA-4, FAM19A4, family with sequence similarity 19 (chemokine (C-C motif)-like), member A4, FLJ25161, TAFA4, TAFA-4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human TAFA4/FAM19A4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human TAFA1, 2, 3, or 5 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only (RUO)