missing translation for 'onlineSavingsMsg'
Learn More

t-Amyl alcohol, MP Biomedicals™

Product Code. 11461516 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461516 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11461516 Supplier MP Biomedicals Supplier No. 0520707780

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 75-85-4
Density 0.809g/mL
Molecular Formula C5H12O
Quantity 100 g
Synonym 2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol
InChI Key MSXVEPNJUHWQHW-UHFFFAOYSA-N
SMILES CCC(C)(C)O
IUPAC Name 2-methylbutan-2-ol
Molecular Weight (g/mol) 88.15
PubChem CID 6405
Formula Weight 88.15
Physical Form Liquid
Chemical Name or Material t-Amyl alcohol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.