Learn More
Thermo Scientific™ T-865 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges
Product Code. 12101690 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
8 x 36 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
12101690 8 x 36 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12101690 Supplier Thermo Scientific™ Supplier No. 51411
  • ONLINE EXCLUSIVE PRICE
    26795.00 EUR
    20560.00 EUR / Pack of 1
    Save 6235.00 EUR
    23% Off
 Restricted Item
In Stock
Perform a variety of applications with the Thermo Scientific™ T-865 Fixed Angle Rotor, which offers eight-place capacity.
Recommended for:
Differential protein fractionation, DNA isolation.
TRUSTED_SUSTAINABILITY
Specifications
Description T-865 Fixed Angle Rotor
Material Titanium
Type Fixed Angle Rotor
Capacity 8 x 36 mL
Max. RCF 429,459 x g
Max. Speed 65,000 rpm
Radius (Metric) 91 mm
Radius (Metric) Min. 38.4 mm
K Factor 51.7
Max. Tube Length 97 mm
Max. Tube Diameter 25 mm
Net Weight 18.3 lb. (8.3 kg)
For Use With (Equipment) Thermo Scientific™ Sorvall™ WX ultraCentrifuges
Angle 23.5°
Avg. RCF 305,340 x g
Min. RCF 181,222 x g