Learn More
CELLPATH SYSTEM II+ MICRO BIOPSY CASSETTE
Description
- Read rates
- Robust and secure closure ensures specimen security
- Designed to improve reagent flow
- Large color range
- Writing surface suitable for marking with a CellMark pen
Specifications
Specifications
| Color | White |
| Material | Loose |
| Product Type | Cassette |