Learn More
Synthetic Sea Water for ASTM 665 / IP 135, Paragon Scientific™
Product Code. 16090420
Ansicht ändern
Click to view available options
Quantity:
300 mL
500 mL
10 x 30 mL
5 L
Packungsgröße:
300mL
5 L
500mL
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
16090420 500 mL 500mL
16070420 10 x 30 mL Pack of 10
16080420 300 mL 300mL
16000430 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16090420 Lieferant Paragon Scientific Lieferanten-Nr. SSWS500
 Restricted Item
Estimated Shipment: 22-09-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
A high quality analytical reagent manufactured and certified compliant for use in ASTM D 665 / IP 135 the Standard Test Method for Rust-Preventing Characteristics of Inhibited Mineral Oil in the Presence of Water.
  • A Synthetic Sea Water Analytical Reagent manufactured in accordance with ASTM D 665 / IP 135.
  • Provided with a certificate which includes an estimation of uncertainty of manufacture.
  • Ensures full compliance with ASTM and IP test method requirements for reagent preparation.
  • Conveniently packaged in single dose or multi dose packs.
  • Cost effective and time saving
  • Certificate of Manufacture & MSDS included
  • 12 month shelf life
Spezifikation
Certifications/Compliance ASTM 665, IP 135
Quantity 500 mL