Learn More
Syngene™ Neutral Field Screen Set for use with IG3 and UG3 systems
Contains a blue screen and a frosted screen. Syngene™ Neutral Field Screen Set is for use with IG3 and UG3 systems.
Brand: Syngene™ GE-NFS
Additional Details : Weight : 1.00000kg
Description
- Corrects for any uneven Illumination by acquiring a neutral-field reference image with the same intensity illumination as the experiment.
- For Epi-UV lighting and transilluminators in Syngene™ Ingenius and Nugenius Systems
Specifications
Neutral Field Screen Set | |
IG3 and UG3 Systems | |
Syngene Ingenius and Nugenius Systems | |
330 x 310 x 15 mm |