Learn More
Pacific Handy Cutter Swivel Holster for S4, S4S and S5
Product Code. 17719047
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17719047 Black Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17719047 Supplier Pacific Handy Cutter Supplier No. UKH423
  • ONLINE EXCLUSIVE PRICE
    92.07 EUR
    78.80 EUR / Pack of 10
    Save 13.27 EUR
    14% Off
Estimated Shipment: 12-01-2027
The UKH-423 Plastic Swivel Holster helps employees retain cutters while increasing productivity and safety.
  • The holster's innovative design acts as an additional safety measure following each use.
  • If left exposed, the holster automatically retracts the blade as soon as the PHC Safety Cutter is placed into the holster to prevent potential injuries.
  • The unique swivel design rotates the holster for the most comfortable positions on the user's hip while sitting or standing.
  • Users also observe noticeable increases in cutter retention and report substantial reductions in replacement costs when utilized in combination with the corresponding PHC Safety Cutter and CL-36 Lanyard.
  • The UKH-423 Holster is specially designed for the S4, S4S and S5 Safety Cutters.
Specifications
For Use With (Equipment) For S4, S4S and S5
Material Plastic
Color Black