Learn More
Pacific Handy Cutter Swivel Holster for QBAR/EZAR
Product Code. 17769047
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17769047 Black Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17769047 Supplier Pacific Handy Cutter Supplier No. UKH675
  • ONLINE EXCLUSIVE PRICE
    78.81 EUR
    67.43 EUR / Pack of 10
    Save 11.38 EUR
    14% Off
Estimated Shipment: 30-12-2026
Practical belt holster made of plastic with fastening clip for belt or clothing. The knife holder can be swiveled.
  • As a result, a comfortable wearing position can be achieved at any time, whether standing or sitting.
  • Suitable for the QBAR and EZAR safety knives.
  • Particularly easy to clean.
Specifications
Product Type Swivel Holster
For Use With (Equipment) QBAR and EZAR Safety Knives
Material Plastic
Color Black