Learn More
3M™ Easy Trap™ Sweep & Dust Sheets
Product Code. 17118710
Change view
Click to view available options
Dimensions (L x W x H):
0.67 x 0.50 x 125 ft.
Unit Size:
1 roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
17118710 0.67 x 0.50 x 125 ft. 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17118710 Supplier 3M™ Easy Trap™ Supplier No. 7100104366
  • ONLINE EXCLUSIVE PRICE
    179.00 EUR
    104.09 EUR / 1 rolls
    Save 74.91 EUR
    41% Off
Estimated Shipment: 14-01-2027
Reduce the risk of bad impressions by trapping up to 8x more dirt, dust, and sand than traditional dust mops!
  • Convenient, disposable sheets mean no laundering
  • Use under any flat floor tool or high duster
  • Sheets are two-sided and come in rolls perforated every 6 in.
  • Low drag reduces worker fatigue
  • Works in wet environments, and on most horizontal surfaces
TRUSTED_SUSTAINABILITY
Specifications
Product Type Duster Sheets
Dimensions (L x W x H) 0.67 x 0.50 x 125 ft.
For Use With (Application) Trapping Dust, Dirt, Sand, and Hair