Learn More
BICASA Suspended Undercounter Cabinet with Hinged Door for Kompatto Workstation
Product Code. 30057538 Shop All BICASA Products
Change view
Click to view available options
Includes:
Left Opening Door
Right Opening Door
Frame Color:
Anthracite
Blue
Red
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Frame Color unitSize
30057538 Left Opening Door Anthracite Each
30057558 Left Opening Door Blue Each
30057537 Left Opening Door Red Each
30057584 Right Opening Door Anthracite Each
30057587 Right Opening Door Blue Each
30057557 Right Opening Door Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30057538 Supplier BICASA Supplier No. KDOLHA
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
Suspended Undercounter cabinet with Hinged Door for Kompatto workstation. Made in Fire-retardant particleboard.
  • For Kompatto Series Workstation
  • Suspended under the countertop
  • Doors: 270° degrees opening - Automatic hold-closing hinges
  • Body width mm 500 + mm 20 of ergonomic anodized and painted aluminum handle
  • Steel sheet shelf (height-adjustable) - Keyable lock as option
  • Fire-retardant particleboard body
  • Light Gray color (2 decorative) - mm 19 thickness
  • Frontal drawer panels and doors with ABS rounded rim
Specifications
Product Type Suspended Undercounter Cabinet
Color Light Gray
Frame Color Anthracite
Material Particleboard
No. of Shelves 1
Includes Left Opening Door
For Use With (Equipment) Kompatto Series Workstation
Dimensions (L x W x H) 600 x (500 + 20) x 530 mm
Width (English) 19.7 in.
Width (Metric) 500 mm