Learn More
Fisherbrand™ SureOne™ Aerosol Barrier Pipette Tips
GREENER_CHOICE
Product Code. 11953466 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.1 to 20 μL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
Tip Style:
Beveled
Extended Length, Micro Point, Low Retention
Micro Point
Micro Point, Extended Length
Color:
Clear
Natural
Unit Size:
Pack of 960
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Tip Style Color unitSize
11953466 10 to 100 μL Beveled Natural Pack of 960
11903466 0.1 to 10 μL Micro Point Natural Pack of 960
11913466 0.1 to 10 μL Micro Point, Extended Length Natural Pack of 960
11977714 0.1 to 20 μL Extended Length, Micro Point, Low Retention Clear Pack of 960
11943466 2 to 20 μL Beveled Natural Pack of 960
11963466 20 to 200 μL Micro Point Natural Pack of 960
11987714 30 to 300 μL Micro Point Clear Pack of 960
11973466 100 to 1000 μL Micro Point Natural Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11953466
  • ONLINE EXCLUSIVE PRICE
    81.46 EUR
    49.20 EUR / Pack of 960
    Save 32.26 EUR
    39% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Choose from a comprehensive line of pipet tips for demanding liquid-handling applications
Universal fit for research grade pipetters.
  • Engineered for increased accuracy and are compatible with single-channel, multichannel, and electronic pipetters
  • DNase-/RNase-free; sterile tips are also pyrogen-free
  • Sterile and clear color
  • Filter eliminates aerosol cross-contamination without sealing or trapping samples
  • Tips allow for noncontaminated sample recovery
  • Not autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 10 to 100 μL
Filtered Filtered
Sterility Sterile
Color Natural
Autoclavable Not Autoclavable
Length (English) 2 in.
Length (Metric) 51 mm
Format Hinged Rack
Tip Style Beveled
Disposable Reusable
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line SureOne
Graduations 10 μL to 100 μL
Show More Show Less
As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.