Learn More
Interscience™ Support for Eppendorf™ pipettors
Product Code. 17806706
Change view
Click to view available options
For Use With (Equipment):
easySpiral, easySpiral Pro (1.5 mL and 2 mL)
easySpiral, easySpiral Pro (5 mL)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17806706 easySpiral, easySpiral Pro (1.5 mL and 2 mL) Each
17272254 easySpiral, easySpiral Pro (5 mL) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17806706 Supplier Interscience™ Supplier No. 413017
  • ONLINE EXCLUSIVE PRICE
    105.00 EUR
    101.17 EUR / Pack of 1
    Save 3.83 EUR
    3% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Support stand for Eppendorf™ pipettors.
  • Supports pipettors of various sizes
Specifications
Type Support Stand
For Use With (Application) Holding Pipettor
For Use With (Equipment) easySpiral, easySpiral Pro (1.5 mL and 2 mL)