Learn More
Interscience™ Support for Eppendorf™ pipettors
Product Code. 17272254
Change view
Click to view available options
For Use With (Equipment):
easySpiral, easySpiral Pro (1.5 mL and 2 mL)
easySpiral, easySpiral Pro (5 mL)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17272254 easySpiral, easySpiral Pro (5 mL) Each
17806706 easySpiral, easySpiral Pro (1.5 mL and 2 mL) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17272254 Supplier Interscience™ Supplier No. 413016
  • ONLINE EXCLUSIVE PRICE
    105.00 EUR
    101.17 EUR / Pack of 1
    Save 3.83 EUR
    3% Off
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
Support stand for Eppendorf™ pipettors.
  • Supports pipettors of various sizes
Specifications
Type Support Stand
For Use With (Application) Holding Pipettor
For Use With (Equipment) easySpiral, easySpiral Pro (5 mL)