Learn More
Epredia™ SuperFrost Gold Plus™ Adhesion Slides
Product Code. 11976299 Shop All Epredia Products
Change view
Click to view available options
No. per Pack:
72/Pk.
Unit Size:
Pack of 72
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Pack Unit Size
11976299 72/Pk. Pack of 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11976299 Supplier Epredia™ Supplier No. K5800AMNZ72
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Epredia™ SuperFrost Plus™ Gold slides are recommended for hard-to-hold tissue samples like bone, brain, breast and skin. They are ideal for special stains, immunocytochemical and in-situ DNA hybridization on these tissue types.
  • No time wasted on slide-coating proceduress
  • No money wasted on expensive adhesives
  • Reliable and uniform coating
  • Minimal sample loss
  • Reliable and safe adhesive action
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 75 mm
Width (Metric) 25 mm
No. per Pack 72/Pk.
Color White
Dimensions (L x W) 75 x 25 mm